Digi-Sense Compression Fitting; Probe Diameter, 3/8"; Brass; 3/8" NPT (M)
Representative image only.
Digi-Sense Item # EW-08539-33
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass
  • Max Temperature (° F)425
  • Max Temperature (° C)218
  • DescriptionCompression Fitting; Probe Diameter, 3/8\"; Brass; 3/8\" NPT (M)
More About this Item
Fittings let you adjust the depth your probe extends into a system for temperature readings. Use fittings for straight-shaft probes and metal tubing only. Tighten compression nut to secure and completely seal probe.
Replacement ferrules let you reuse a compression fitting when replacing a worn-out probe. Simply disassemble fitting, discard probe and old ferrule, install replacement ferrule over probe sheath, and retighten fitting.