Digi-Sense Cord Adapter From IEC 60320 to SEV1011 (Switzerland Plug), 12" L; 10A, 240V
Representative image only.
Glas-Col Item # EW-89800-26
  • Switzerland specific female plug
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)240
  • DescriptionCord Adapter From IEC 60320 to SEV1011 (Switzerland Plug), 12\" L; 10A, 240V
  • Warranty1 year
1 Year Warranty
Key Features
  • Switzerland specific female plug
  • 1 year warranty
More About this Item
Choose an interchangeable cord/receptacle assembly for your 230 VAC temperature controller. Each 12" (30.5 cm) cord set features a country-specific female plug on one end and an IEC 60320 C19 male plug on the other end rated for 10 A at 240 VAC.