Cole-Parmer Essentials Dry Block/Incubator Liquid-In-Glass Thermometer; 20 to 100C, PFA Safety Coated, 76mm Immersion, Organic Liquid Fill
Representative image only.
Cole-Parmer Essentials Item # EW-08006-23
Replace mercury thermometers for a specific application with the same accuracy without any of the hazards
  • Non-mercury liquid
  • 5 year warranty
PRODUCT OPTIONS
$79.60USD / Each
$219.50 - $292.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")12
  • Min Temperature (° C)20
  • Max Temperature (° C)100
  • Accuracy±1.0°C
  • Division (° C)1
  • Thermometer FillRed Spirit
  • Overall Length (in)12
  • Overall Length (mm)300
  • Immersion Depth (mm)76
  • Stem MaterialPFA-Coated White-Back Glass
  • DescriptionDry Block/Incubator Liquid-In-Glass Thermometer; 20 to 100C, PFA Safety Coated, 76mm Immersion, Organic Liquid Fill
  • Warranty5 years
5 Year Warranty
Key Features
  • Non-mercury liquid
  • 5 year warranty
More About this Item
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PFA coated for added safety which may cause a slight change in accuracy up to 1 division.
Dry block thermometers - designed for use in all standard heating blocks and water baths with most having arrows indicating critical temperatures.