Digi-Sense Full Immrsn PRT, 0.125" x 2" , -196~180C, 5-Pin Din
Representative image only.
Digi-Sense Item # EW-90452-11
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5606
  • 1 year warranty
PRODUCT OPTIONS
$702.00USD / Each
$520.00 - $593.00USD / Each
for an additional
$520.00USD / Each
for an additional
$593.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeFull Immersion PRT/RTD
  • Min Temperature (° F)-320.8
  • Max Temperature (° F)356
  • Min Temperature (° C)-196
  • Max Temperature (° C)180
  • Temperature Accuracy±0.05°C and ±0.04°C/100 hr
  • Response Time (Sec)4
  • Connector Type5-Pin DIN
  • Sheath Length (in)2
  • Sheath Diameter (in)0.125
  • DescriptionFull Immrsn PRT, 0.125\" x 2\" , -196~180C, 5-Pin Din
  • Warranty1 year
1 Year Warranty
Key Features
  • Equivalent to Fluke Calibration / Hart Scientific Model 5606
  • 1 year warranty
More About this Item
AM1612 and AM1642 Series—Full-immersion PRT/RTD probes: Designed to perform in extreme environments where both the transition junction and the lead wires are required to withstand temperatures covering the entire operating range of the probe. Applications include calibration or validation of sensors used in laboratory or biology freezers, walk-in refrigerators, autoclaves, ovens, stability test chambers, furnaces, and incubators. These probes feature short term stability of 20 mK at TPW and long term drift of 40 mK at Triple Point of Water (TPW). The AM1612 has a temperature range of -196 to 180ºC (-320.8 to 356ºF)