Digi-Sense Incubator Verification Thermometer; 25 to 45C
Representative image only.
Digi-Sense Item # EW-08077-34
Enclosed bottle ensures accuracy and is ideal for use in environmental chambers
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • NIST- traceable statement of accuracy included
  • 5 year warranty
PRODUCT OPTIONS
$78.60USD / Each
$145.50 - $219.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")7 1/2
  • Min Temperature (° C)25
  • Max Temperature (° C)45
  • Accuracy±0.4°C
  • Division (° C)0.1
  • Thermometer FillBlue Spirit
  • Bottle FillGlass Beads
  • Overall Length (in)7.5
  • Overall Length (mm)190
  • Stem MaterialWhite-Back Glass
  • DescriptionIncubator Verification Thermometer; 25 to 45C
  • Warranty5 years
5 Year Warranty
Key Features
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • NIST- traceable statement of accuracy included
  • 5 year warranty
More About this Item
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. All thermometers are supplied in a glass bead filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.