Cole-Parmer Irreversible 1-Point Square Temperature Label, 230F/110C; 50/Pk
Representative image only.
Cole-Parmer Item # EW-90309-35
Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
  • 1 year warranty
PRODUCT OPTIONS
$33.60USD / Pkg of 50
BUY MORE & SAVE
  • 1-4$33.60USD / Pkg of 50
  • 5-9$30.24USD / Pkg of 50
  • 10-19$28.56USD / Pkg of 50
  • 20+$26.88USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators1
  • Temperature Points (° F)230
  • Temperature Points (° C)110
  • Initial colorWhite
  • First color changeBlack
  • Height (in)1/2
  • Width (in)1/2
  • DescriptionIrreversible 1-Point Square Temperature Label, 230F/110C; 50/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
  • 1 year warranty
More About this Item
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F. Shelf life: 1 year after date of purchase.