Digi-Sense Irreversible 8 and 9-Point Temperature Label Kit , 5 of Each 5 Types
Representative image only.
Digi-Sense Item # EW-08068-90
Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
PRODUCT OPTIONS
$58.50USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$58.50USD / Pkg of 25
  • 10-24$52.65USD / Pkg of 25
  • 25-49$49.73USD / Pkg of 25
  • 50+$46.80USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators9
  • Max temperature (° F)500
  • Max temperature (° C)260
  • Height (in)2
  • Width (in)3 3/4
  • DescriptionIrreversible 8 and 9-Point Temperature Label Kit , 5 of Each 5 Types
Key Features
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
More About this Item
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F. Shelf life: 1 year after date of purchase.
Contains five labels of each of the five types of indicators. Product numbers: 08068-20, 08068-22, 08068-24, 08068-26, and 08068-28.