Digi-Sense Liquid-In-Glass Oven Thermometer; 20 to 130C, Organic Liquid Fill, Total Immersion

Representative image only.
Digi-Sense Item # EW-08006-43
Replace mercury thermometers for a specific application with the same accuracy without the any hazards
  • Non-mercury liquid

PRODUCT OPTIONS

$26.25USD / Each

$198.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Monitor your critical environments

SPECS & DESCRIPTION

Specifications & Description

  • Length (")5 19/64
  • Min Temperature (° C)20
  • Max Temperature (° C)130
  • Accuracy±2°C
  • Division (° C)2
  • Thermometer FillRed Spirit
  • Overall Length (in)5.3
  • Overall Length (mm)135
  • Immersion Depth (mm)135
  • Stem MaterialWhite-Back Glass
  • DescriptionLiquid-In-Glass Oven Thermometer; 20 to 130C, Organic Liquid Fill, Total Immersion

Key Features

  • Non-mercury liquid

More About this Item

These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PTFE coated for added safety which may cause a slight change in accuracy up to 1 division.

Oven thermometers - designed for high temperature applications and feature a round glass swell to hold the thermometer in place

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat