Digi-Sense Panel-Mount Standard Connector, Type-T, Female, Rectangular, 1/Ea
Representative image only.
Digi-Sense Item # EW-18527-32
Round or rectangular shape to fit your instrument panel
  • Glass-filled nylon construction provides rigidity, strength, and surface hardness
  • Molded barriers prevent short circuit
  • 6 month warranty
PRODUCT OPTIONS
$11.50USD / Each
BUY MORE & SAVE
  • 1-1$11.50USD / Each
  • 2-9$10.35USD / Each
  • 10+$9.78USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connector TypeJack
  • Special InformationGlass-filled nylon body
  • Max Temperature (° F)392
  • Max Temperature (° C)200
  • DescriptionPanel-Mount Standard Connector, Type-T, Female, Rectangular, 1/Ea
  • Warranty6 month
6 Month Warranty
Key Features
  • Glass-filled nylon construction provides rigidity, strength, and surface hardness
  • Molded barriers prevent short circuit
  • 6 month warranty
More About this Item
Mount connectors to your panel using the two screws located on the front of the jack. Each connector is ANSI color-coded by thermocouple type: black (J), yellow (K), or blue (T). Choose between flat-bladed miniconnectors and round-pinned standard connectors.