Digi-Sense Precision Liquid-In-Glass Thermometer; 30 to 124F, Total Immersion, Organic Liquid Fill
Representative image only.
Digi-Sense Item # EW-90260-30
Scales are permanently fused on to the glass and won’t wear off
  • Nonmercury blue spirit liquid
  • Statement of accuracy included
PRODUCT OPTIONS
$83.60USD / Each
$219.50 - $292.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Stay connected — Monitor your critical environments
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)30
  • Length (\")17 29/32
  • Max Temperature (° F)124
  • Accuracy±2.0°F
  • Division (° F)0.2
  • Thermometer FillBlue Spirit
  • Overall Length (in)17.9
  • Overall Length (mm)455
  • Immersion Depth (mm)455
  • Stem MaterialWhite-Back Glass
  • DescriptionPrecision Liquid-In-Glass Thermometer; 30 to 124F, Total Immersion, Organic Liquid Fill
5 Year Warranty
Key Features
  • Nonmercury blue spirit liquid
  • Statement of accuracy included
More About this Item
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Fractional-degree thermometers offer higher precision. Choose from partial immersion or total immersion thermometers.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.