Cole-Parmer Reversible 48-Point Vertical Adhesive Temperature Label, 154-184F; 25/Pk
Representative image only.
Cole-Parmer Item # EW-09100-07
Each style covers a wide range of temperatures, for use in many different application
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 2 year warranty
PRODUCT OPTIONS
$41.10USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators48
  • Max temperature (° F)184
  • Max temperature (° C)16
  • Temperature Points (° F)154, 156, 158, 160, 162, 164, 166, 168, 170, 172, 174, 176, 178, 180, 182, 184
  • Temperature Points (° C)68, 69, 70, 71, 72, 73, 74, 75, 76, 77, 78, 79, 80, 81, 82, 83
  • Initial colorBlack
  • First color changeGreen (actual temperature); Brown (above actual); Blue (below actual)
  • Second color changeBlack
  • Height (in)5
  • Width (in)1/2
  • DescriptionReversible 48-Point Vertical Adhesive Temperature Label, 154-184F; 25/Pk
  • Warranty2 year
Two year warranty
Key Features
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 2 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.