Cole-Parmer Reusable Low-Temperature Labels, Cooler, 34 to 50F (1 to 10C); 10/Pk
Representative image only.
Cole-Parmer Item # EW-90308-90
Ideal for use in refrigerator, freezer and cooler applications
  • Nontoxic nonhazardous temperature-measuring alternative
  • 1 year warranty
PRODUCT OPTIONS
$33.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators1
  • Max temperature (° F)50
  • Max temperature (° C)10
  • Temperature Points (° F)34 to 50
  • Temperature Points (° C)1 to 10
  • Height (in)5 3/8
  • Width (in)1 1/2
  • DescriptionReusable Low-Temperature Labels, Cooler, 34 to 50F (1 to 10C); 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Nontoxic nonhazardous temperature-measuring alternative
  • 1 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring low temperatures. Indicating strips use liquid crystal technology to display the current temperature range at an economical cost. Shelf life: 1 year after date of purchase.