Cole-Parmer Reversible 13-Point Horizontal Temperature Label, 36-48C / 97-118F; 10/Pk

Representative image only.
Cole-Parmer Item # EW-09035-47
Each style covers a wide range of temperatures, for use in many different application
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty

PRODUCT OPTIONS

$29.30USD / Pkg of 10

BUY MORE & SAVE

  • 1-4$29.30USD / Pkg of 10
  • 5-9$26.37USD / Pkg of 10
  • 10-19$24.91USD / Pkg of 10
  • 20+$23.44USD / Pkg of 10

SPECS & DESCRIPTION

Specifications & Description

  • Number of Temperature Indicators13
  • Max temperature (° F)118
  • Max temperature (° C)48
  • Temperature Points (° F)97, 99, 100, 102, 104, 106, 108, 109, 111, 113, 115, 116, 118
  • Temperature Points (° C)36, 37, 38, 39, 40, 41, 42, 43, 44, 45, 46, 47, 48
  • Initial colorBlack
  • First color changeGreen
  • Second color changeBlue
  • Length (in)39/64
  • Depth (in)5 13/64
  • DescriptionReversible 13-Point Horizontal Temperature Label, 36-48C / 97-118F; 10/Pk
  • Warranty1 year
1 Year Warranty

Key Features

  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty

More About this Item

These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat