Digi-Sense RTD Probe with 4 to 20 mA Output, 0 to 400°F, 12" Immersion Depth
Representative image only.
Digi-Sense Item # EW-32884-04
  • Hermetically sealed electronics make these probes ideal for food, dairy, and pharmaceutical applications
  • Use optional software to rescale or field calibrate probes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeRTD
  • Min Temperature (° F)-40
  • Max Temperature (° F)158
  • Min Temperature (° C)-40
  • Max Temperature (° C)70
  • Connector TypeStripped Leads
  • DescriptionRTD Probe with 4 to 20 mA Output, 0 to 400°F, 12\" Immersion Depth
  • Warranty1 year
1 Year Warranty
Key Features
  • Hermetically sealed electronics make these probes ideal for food, dairy, and pharmaceutical applications
  • Use optional software to rescale or field calibrate probes
  • 1 year warranty
More About this Item
These RTD probes feature mini transmitters that are integrated into the sensors and hermetically sealed to protect them from fluid and harsh washdowns. Probes come factory calibrated to measuring range. Fittings are adjustable along the immersion depth of the sensor. Order the calibration software separately.