Digi-Sense RTD Stranded Wire, 28-Gauge, TFE/FEP Insulation, Duplex, 100-ft Spool
Representative image only.
Digi-Sense Item # EW-08541-41
Extend the measuring distance of your probes
  • 100-foot spool of RTD wire- cut to any lenght needed!
SPECS & DESCRIPTION
Specifications & Description
  • Wire Gauge (Ga)28
  • Inner InsulationPolytetrafluoroethylene/Fluorinated ethylene propylene
  • Outer InsulationPolytetrafluoroethylene/Fluorinated ethylene propylene
  • Stranded / SolidStranded
  • Special InformationDuplex / 6 wires
  • Min Temperature (° F)-328
  • Max Temperature (° F)400
  • Min Temperature (° C)-200
  • Max Temperature (° C)204
  • Length (ft)100
  • DescriptionRTD Stranded Wire, 28-Gauge, TFE/FEP Insulation, Duplex, 100-ft Spool
Key Features
  • 100-foot spool of RTD wire- cut to any lenght needed!
More About this Item
Three-strand RTD wire features excellent resistance to abrasion, chemicals, and moisture. Each silver-plated copper strad is color-coded with fluorinated ethylene propylene (FEP). The three starnds are twisted to increase flexibility and minimize electromagnetic noise, then jacketed in white FEP. The FEP insulation provides temperature resistance up to 400°F, and chemical resistance to solvents, acids, and oils. Additionally, the insulation protects the wire from thermal aging, while maintaining its strength and flexibility.