Digi-Sense Sec. Ref. PRT, 0.25" x 20" , -200~670C, Bare Wire
Representative image only.
Digi-Sense Item # EW-90452-44
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5609
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeSecondary Reference PRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)1238
  • Min Temperature (° C)-200
  • Max Temperature (° C)670
  • Temperature Accuracy±0.015°C and ±0.01°C/100 hr
  • Response Time (Sec)9
  • Connector TypeStripped Wire
  • Sheath Length (in)20
  • Sheath Diameter (in)0.25
  • DescriptionSec. Ref. PRT, 0.25\" x 20\" , -200~670C, Bare Wire
  • Warranty1 year
1 Year Warranty
Key Features
  • Equivalent to Fluke Calibration / Hart Scientific Model 5609
  • 1 year warranty
More About this Item
AM1750 Series—Secondary reference PRT/RTD probes: These probes are the workhorse of laboratories. With a calibration uncertainty at 0°C of just ±0.015°C, these probes fully meet ITS-90 criteria for reference thermometers fully. The unique design achieves an excellent balance between hysteresis effect and mechanical/thermal shocks. Both series boast very low short term stability of 7 mK at TPW. The AM1750 has a temperature range of -200 to 670°C,
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.