Digi-Sense Sec. SPRT, 0.25" x 12" , -200~670C, 5-Pin Din SKU 9045259

Representative image only.
Digi-Sense Item # EW-90452-59
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5628

PRODUCT OPTIONS

$1,678.00USD / Each

$449.00USD / Each

for an additional
$449.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Probe TypeSecondary Reference PRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)1238
  • Min Temperature (° C)-200
  • Max Temperature (° C)670
  • Temperature Accuracy±0.006°C and ±0.004°C/100 hr
  • Response Time (Sec)9
  • Connector Type5-Pin DIN
  • Sheath Length (in)12
  • Sheath Diameter (in)0.25
  • DescriptionSec. SPRT, 0.25" x 12" , -200~670C, 5-Pin Din

Key Features

  • Equivalent to Fluke Calibration / Hart Scientific Model 5628

More About this Item

AM1762 Series—Secondary standard SPRT/RTD probes: These probes fill the gap between affordable, but temperature limited secondary PRTs and more expensive, yet highly accurate SPRTs. The unique design makes them very stable both short term (±2 mK at TPW) and long term (<19 mK typical at TPW after 500 hours) accuracy is ±6 mK at TPW . These high performance probes fully meet ITS-90 criteria for reference thermometers and are an excellent choices for reference standards for metrology labs of various levels. The Am1762 is nominally 25.5 ohm

Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat