Digi-Sense Sec. SPRT, 0.25" x 12" , -200~670C, Spade SKU 9045249
Representative image only.
Digi-Sense Item # EW-90452-49
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5626
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeSecondary Reference PRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)1238
  • Min Temperature (° C)-200
  • Max Temperature (° C)670
  • Temperature Accuracy±0.006°C and ±0.004°C/100 hr
  • Response Time (Sec)9
  • Connector TypeSpade Lugs
  • Sheath Length (in)12
  • Sheath Diameter (in)0.25
  • DescriptionSec. SPRT, 0.25\" x 12\" , -200~670C, Spade
  • Warranty1 year
1 Year Warranty
Key Features
  • Equivalent to Fluke Calibration / Hart Scientific Model 5626
  • 1 year warranty
More About this Item
AM1760 Series—Secondary standard SPRT/RTD probes: These probes fill the gap between affordable, but temperature limited secondary PRTs and more expensive, yet highly accurate SPRTs. The unique design makes them very stable both short term (±2 mK at TPW) and long term (<19 mK typical at TPW after 500 hours) accuracy is ±6 mK at TPW . These high performance probes fully meet ITS-90 criteria for reference thermometers and are an excellent choices for reference standards for metrology labs of various levels. The AM1760 is nominally 100 ohm
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.