Digi-Sense Solar-Powered Adjustable-Angle Celsius Thermometer; 6" L
Representative image only.
Digi-Sense Item # EW-90131-06
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° C)-50
  • Max Temperature (° C)150
  • Division (° C)0.1
  • Dial / Case Material304 Stainless Steel
  • Stem Material304 Stainless Steel
  • DescriptionSolar-Powered Adjustable-Angle Celsius Thermometer; 6\" L
  • Warranty1 year
1 Year WarrantyNSF international certified
Key Features
  • 1 year warranty
More About this Item
View readings from almost any angle by simply adjusting the angle of the head. Three-inch diameter adjustable head has an easy-to-read LCD. Requiring no batteries these thermometers are ideal in pilot plant or research and development applications where exact readings are required. The safety circuit prevents false readings.
Each thermometer is housed in a weather-resistant 304 stainless steel (SS) case. Stem is 304 SS. An acrylic lens protects the face. To further protect thermometer order a thermowell from page 1052. Simply match the stem length to the probe depth "A" for the thermowell.