Digi-Sense Temperature Probe, Type K, 10 M, with Mini-Connector, ector
Representative image only.
Digi-Sense Item # EW-08879-84
  • Special use thermocouple probes
  • 6 month warranty
PRODUCT OPTIONS
$42.31USD / Each
$47.02USD / Each
$197.00 - $270.00USD / Each
for an additional
$197.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeK
  • Min Temperature (° F)-148
  • Max Temperature (° F)482
  • Min Temperature (° C)-100
  • Max Temperature (° C)250
  • Connector TypeMini Connector
  • DescriptionTemperature Probe, Type K, 10 M, with Mini-Connector
  • Warranty6 month
6 Month Warranty
Key Features
  • Special use thermocouple probes