Digi-Sense Temperature Transmitter, 3-Wire 100 Ohm Pt RTD, -100 to 100F; 2.25" x 1.5"
Representative image only.
Digi-Sense Item # EW-94700-44
$238.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Control Input RTD100 Ω Pt, 3-wire
  • Output4 to 20 mA
  • Min Temperature (° F)-100
  • Max Temperature (° F)100
  • Loop Power Supply13 to 40 VDC
  • DescriptionTemperature Transmitter, 3-Wire 100 Ohm Pt RTD, -100 to 100F; 2.25" x 1.5"
More About this Item
Thermocouple and RTD Transmitters