Digi-Sense Thermocouple Probe Wire, Type T, 24-Gauge, Glass Braid with SS Overbraid, 100ft
Representative image only.
Digi-Sense Item # EW-08541-37
Make your own extension cables
PRODUCT OPTIONS
$210.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialFiberglass
  • Wire Gauge (Ga)24
  • Inner InsulationFiberglass Wrap
  • Outer InsulationFiberglass Wrap
  • Stranded / SolidSolid
  • OverbraidYes
  • Min Temperature (° F)-100
  • Max Temperature (° F)900
  • Min Temperature (° C)-73
  • Max Temperature (° C)482
  • Length (ft)100
  • DescriptionThermocouple Probe Wire, Type T, 24-Gauge, Glass Braid with SS Overbraid, 100ft
More About this Item
Wire comes in 24-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.