Digi-Sense Thermocouple Wire, Type-J, 20-Gauge, Polyvinyl Insulation, 1000-ft Spool
Representative image only.
Digi-Sense Item # EW-08542-42
Make your own extension cables
PRODUCT OPTIONS
$480.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyvinyl chloride (PVC)
  • Wire Gauge (Ga)20
  • Inner InsulationPolyvinyl chloride (PVC)
  • Outer InsulationPolyvinyl chloride (PVC)
  • Stranded / SolidSolid
  • OverbraidNo
  • Special InformationWith BoPET shied and drain wire
  • Min Temperature (° F)-20
  • Max Temperature (° F)221
  • Min Temperature (° C)-28
  • Max Temperature (° C)105
  • Length (ft)1000
  • DescriptionThermocouple Wire, Type-J, 20-Gauge, Polyvinyl Insulation, 1000-ft Spool
More About this Item
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.