Digi-Sense Type K Thermocouple Inconel Probe Quick DisConnector, Dual with Std-Connector, 12" L, .188 Dia, Grounded Junction
Representative image only.
Digi-Sense Item # EW-18520-87
Designed for reading two temperatures at one time
  • Ideal for continuous monitoring
  • Designed with ICONEL 600 sheath
  • Response Time: 1.2 - 5 seconds or greater
  • Includes a stand alone female connector for instalation to a thermocouple extension cable
  • Each probe comes with a color coded ANSI dual standard connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")12
  • Probe TypeK
  • Min Temperature (° F)-250
  • Max Temperature (° F)1650
  • Min Temperature (° C)-150
  • Max Temperature (° C)1100
  • Temperature Accuracy±4.0°F (2.2°C) or ±0.4% reading above 32° (0°C); ±4.0% of reading below 32°F (0°C)
  • JunctionGrounded
  • Connector TypeStandard Connector, Dual
  • Sheath materialInconel®
  • Sheath Length (in)12
  • Sheath Diameter (in)0.1875
  • DescriptionType K Thermocouple Probe Probe Quick Disconnect Dual with Std Con, 12\" L, .188 Dia, Grn Jct
  • Warranty6 month
6 Month Warranty
Key Features
  • Ideal for continuous monitoring
  • Designed with ICONEL 600 sheath
  • Response Time: 1.2 - 5 seconds or greater
  • Includes a stand alone female connector for instalation to a thermocouple extension cable
  • Each probe comes with a color coded ANSI dual standard connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty
More About this Item
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200