Digi-Sense Type T Disc Surface Thermocouple Probe
Representative image only.
Digi-Sense Item # EW-08506-80
  • Ideal for checking small surfaces
  • Grounded junction for safety from contaminants
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeK
  • Min Temperature (° F)-418
  • Max Temperature (° F)194
  • Min Temperature (° C)-250
  • Max Temperature (° C)90
  • Time Constant0.15 second
  • Response Time (Sec)0.75
  • JunctionGrounded
  • Connector TypeMini Connector
  • Sheath Diameter (in)0.3125
  • DescriptionType T Disc Surface Thermocouple Probe
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for checking small surfaces
  • Grounded junction for safety from contaminants
  • 1 year warranty
More About this Item
Tape vinyl-insulated 10-kt gold-plated disc sensor onto any flat surface.