Digi-Sense Type T Fine -Gauge PTFE-Insulated Thermocouple Probe
Representative image only.
Digi-Sense Item # EW-08506-75
Ideal for chemical resistant applications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeT
  • Min Temperature (° F)-418
  • Max Temperature (° F)302
  • Min Temperature (° C)-250
  • Max Temperature (° C)150
  • Time Constant0.1 second
  • Response Time (Sec)0.5
  • JunctionUngrounded
  • Connector TypeMini Connector
  • Sheath materialPTFE Insulated Wire
  • Sheath Diameter (in)0.024
  • DescriptionType T Fine-Gauge PTFE-Insulated Thermocouple Probe
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded miniconnectors: type T—blue. This is a fine-gauge PTFE-insulated probe, with a 0.025" outer diameter and is 3-ft long. This is a tissue implantable microprobe, ideal for semisolids.