Digi-Sense Universal Industrial Thermocouple Probes Type K, 24" Length
Representative image only.
Digi-Sense Item # EW-93830-77
A low cost alternative to thermowell assemblies
  • 1 year warranty
PRODUCT OPTIONS
$271.75USD / Each
$197.00 - $270.00USD / Each
for an additional
$197.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeK
  • Min Temperature (° F)0
  • Max Temperature (° F)2000
  • Min Temperature (° C)-17
  • Max Temperature (° C)1093
  • JunctionGrounded
  • Connector Type1/2" NPT(M) x 3/4" NPT(F) conduit
  • Sheath material446 Stainless Steel
  • Sheath Length (in)24
  • DescriptionUniversal Industrial Thermocouple Probes Type K, 24" Length
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The Uniwell eliminates the need for thermowells and spring loaded probes. With a unique adjustable feature that allows the probe to be customized to the required insertion length, through the use of an adjustable ½” NPT compression fitting. Also designed to be directly inserted into the medium being measured without the use of a thermowell. This allows for ease of installation where a thermowell is not required. The probes conductor is made of 14 gauge wire and is encased with Magnesium Oxide insulation, and has a 446 stainless steel heavy wall sheath producing an increased life span. Featuring a standard size aluminum protection head with a gasketed cap to seal out dust and moisture and houses a ceramic terminal block. Head has a 3/4" NPT(F) opening on the side for easy connection to