Digi-Vac TRAC/VAC Visual Light Bar Vacuum Meter; milliTorr

Representative image only.
Digi-Vac Mfr # TRACVAC – Item # EW-38436-10
Progressive, dynamic LED bar graph alerts you to vacuum trends and set points at a glance
  • Trending feature visually illustrates vacuum rate of change
  • Set point indicator function flashes LEDs when the vacuum system surpasses a set point
  • Rugged thermocouple sensor and vinyl-clad metal housing are suitable for harsh environments
  • Bright green LED light bar can be easily seen from across a room

$712.50USD / Each

$184.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±15% of reading up to 2000 micron, ±50% of reading above 2000 micron
  • Max Vacuum (" Hg)30
  • Min Vacuum (" Hg)0
  • Process Connection1/8" NPT(M) or 1/4" flare
  • Unit Of Measurementmicron
  • DisplayGreen LED bar graph, 30 segments
  • Battery Type100 to 240 VAC, 50/60 Hz
  • Media CompatibilityDry air and nitrogen
  • Depth (in)5 1/2
  • Height (in)1 1/2
  • Width (in)4 1/4
  • DescriptionVisual Light Bar Vacuum Meter; milliTorr

Includes

thermocouple vacuum sensor with 10-ft (3-m) cable and power cord.

Key Features

  • Trending feature visually illustrates vacuum rate of change
  • Set point indicator function flashes LEDs when the vacuum system surpasses a set point
  • Rugged thermocouple sensor and vinyl-clad metal housing are suitable for harsh environments
  • Bright green LED light bar can be easily seen from across a room

More About this Item

Designed for higher accuracy at higher vacuum (below 2000 micron) levels, the TRAC/VAC vacuum meter is well suited for critical laboratory and industrial applications. Field calibration allows you to adjust the zero point for greater accuracy, or when the vacuum system contains a gas other than nitrogen or air.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat