Double Open End Carbon-Impregnated Paper Cartridge; 5um, 10"L, 1/Pk
Representative image only.
Item # EW-01509-25
Ideal for chemically neutral waters with lower chlorine concentrations
PRODUCT OPTIONS
$15.20USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaActivated Carbon
  • Micron Rating5
  • Flow Rate5 GPM at 4.0 psid
  • Filter Length (in)10
  • Max Temperature (° F)125
  • Max Temperature (° C)51.7
  • DescriptionDouble Open End Carbon-Impregnated Paper Cartridge; 5um, 10"L, 1/Pk
More About this Item
This cellulose cartridge removes organic tastes/odors and chlorine, plus reduces sediment. Suitable to 125°>F (52°>C).
Treat approximately 2500 gallons at 1 GPM for chlorine (filtration efficiency and chlorine reduction are reduced at higher flow rates; chlorine capacity based on 75% reduction using 2 ppm free chlorine feed concentration at 68°F (20°C)). Meets NSF Standard 42 for materials.