Double open end silver/carbon cartridge; 25 µm, 10"L
Representative image only.
Item # EW-01508-91
$92.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaActivated Carbon
  • Micron Rating25
  • Flow Rate1 GPM
  • Filter Length (in)25
  • Max Temperature (° F)90
  • Max Temperature (° C)32
  • DescriptionDouble open end silver/carbon cartridge; 25 µm, 10"L
More About this Item
Silver/Carbon Cartridge has silver impregnated in activated carbon to inhibit growth of bacteria. Removes solids, organic compounds, chlorine, tastes, and odors. For cold water only. Produces 3500 gallons prior to exhaustion. Max temperature is 77°F (25°C); max differential pressure is 10 psid.
Note: Cartridge does not work with DOE housing models 01508-40, -41, and -42.
* Psid refers to pounds per square inch pressure drop through the filter system