Double open end spun PP progressive-density cartridge; 1 µm, 10 inch length
Representative image only.
Item # EW-01509-14
Spun gradient-density construction provides greater dirt holding capacity
PRODUCT OPTIONS
$6.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating1
  • Flow Rate5 GPM at 0.6 psid
  • Filter Length (in)10
  • Max Temperature (° F)145
  • Max Temperature (° C)62.8
  • DescriptionDouble open end spun PP progressive-density cartridge; 1 µm, 10 inch length
More About this Item
Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust, and scale. Polypropylene construction is resistant to chemical and bacterial attack. Suitable to 145°F (63°C). Note: For optimal performance, change cartridge at 25 psid.