Drager CMS Analyzer Carbon dioxide, 1 to 20%
Representative image only.
Drager Mfr # 6406210 – Item # EW-86513-18
Eliminate user error for a consistent measurement every time
  • Digital display of gas concentration eliminates ambiguous results
  • Internal recorder stores up to 50 measurements with time/date stamp
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredCarbon dioxide
  • Range1 to 20%
  • DescriptionCarbon dioxide, 1 to 20%
  • Warranty2 year
Two year warranty
Key Features
  • Digital display of gas concentration eliminates ambiguous results
  • Internal recorder stores up to 50 measurements with time/date stamp
  • 2 year warranty
More About this Item
Just load a CMS gas specific chip into the bottom port of the analyzer. Easy-to-use, three-step slide switch allows analyzer to take measurement. Gas concentration is displayed in seconds with digital readout. Operates on four AA batteries (included). Chips contain 10 tests each and are sold separately.