Drager Diffusion Tube, Butadiene 10/a-D, 10 to 300 ppm at 1 hour
Representative image only.
Drager Mfr # 8101161 – Item # EW-86515-53
Direct read tubes identify workers exposure to chemicals
  • Read exposure levels easily but he markings on the side of the tube
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredButadiene 10/a-D
  • Range1 hr: 10 to 300 ppm, 4 hr: 1.3 to 40 ppm
  • DescriptionDiffusion Tube, Butadiene 10/a-D, 10 to 300 ppm at 1 hour
  • Warranty2 year
Two year warranty
Key Features
  • Read exposure levels easily but he markings on the side of the tube
  • 2 year warranty
More About this Item
Direct-reading diffusion tubes are used specifically for personal exposure monitoring. These tubes do not require the use of a gas detector pump because the contaminant molecules pass into the tube by diffusion processes and in this manner are brought into contact with the reagent systems. Exposure levels are easily read by the markings on the side of the tube.