Drager Gas Detection Tubes, Ethylene, 0.2 To 5 ppm, 10/box (5 tests/box)
Representative image only.
Drager Mfr # 8101331 – Item # EW-86514-54
Easy, one-handed operation
  • Wider diameter tubes with well-spaced graduation marks are easier to read
  • 2 year warranty
PRODUCT OPTIONS
$136.00USD / of 10
$160.00USD / of 10
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredEthylene
  • Range0.2 to 5 ppm
  • DescriptionGas Detection Tubes, Ethylene, 0.2 To 5 ppm, 10/box (5 tests/box)
  • Warranty2 year
Two year warranty
Key Features
  • Wider diameter tubes with well-spaced graduation marks are easier to read
  • 2 year warranty
More About this Item
The accuro® gas detector pump is a manually operated
bellows pump that draws a calibrated 100-mL sample
through the short-term Dräger gas detection tubes
(order pump separately). Provides fast, accurate, reliable
results in just seconds.