Drager PAC 3500 Single-Gas Monitors, CO, 0 to 500 ppm

Representative image only.
Drager Mfr # 4543957 8322001 – Item # EW-86515-03
Three alarms functions – audible, visual, and vibration – make alarm conditions clear
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Gases MeasuredCO
  • Min CO Range (ppm)0
  • Max CO Range (ppm)500
  • CO Resolution (ppm)1
  • CO Alarm Set Points35 ppm (low); 50 ppm (high)
  • Sensor Life2 years
  • Alarm90 dB (audible), visual, vibration
  • Battery TypeNonreplaceable lithium batteries
  • Height (in)2 1/2
  • Width (in)3 1/4
  • Depth (in)1
  • Height (cm)6.5
  • Width (cm)8.5
  • Depth (cm)2.5
  • DescriptionSingle-Gas Monitors, CO, 0 to 500 ppm
  • Warranty2 year
Two year warrantyConformite EuropeenneIP 65

Key Features

  • 2 year warranty

More About this Item

The monitor features a robust, rubber-coated weatherproof polymer housing rated to IP65 that will stand up to the typical abuse in an industrial environment. Easily operated using just two buttons. Its small size makes it comfortable and easy to wear all day on a belt, pocket, or lapel. A loud audible alarm, two bright visual alarms, and strong vibrating alarm alert you to hazardous concentrations of gas. Built-in event logger stores up to 60 alarm activations or other events.

The Pac 3500 is a maintenance-free detector with a two-year sensor life.
Approval: UL and cUL classified intrinsically safe in Class I, Div. 1, Groups A, B, C, D, E, F, G T4; ATEX; CE
What's included: Pocket/belt clip and batteries.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat