Drager Tubes, Carbon Monoxide, 0.1 to 7%, 10/box

Representative image only.
Drager Mfr # CH29901 – Item # EW-86514-28
Easy, one-handed operation
  • Wider diameter tubes with well-spaced graduation marks are easier to read
  • 2 year warranty

PRODUCT OPTIONS

$127.00USD / of 10

SPECS & DESCRIPTION

Specifications & Description

  • Gas MeasuredCarbon monoxide
  • Range0.1 to 7%
  • TLV (ppm)25
  • DescriptionTubes, Carbon Monoxide, 0.1 to 7%, 10/box
  • Warranty2 year
Two year warranty

Key Features

  • Wider diameter tubes with well-spaced graduation marks are easier to read
  • 2 year warranty

More About this Item

The accuro® gas detector pump is a manually operated
bellows pump that draws a calibrated 100-mL sample
through the short-term Dräger gas detection tubes
(order pump separately). Provides fast, accurate, reliable
results in just seconds.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat