DuPont IC182BWH00250C Coveralls, x-large, cleaned and individually wrapped
Representative image only.
DuPont Mfr # IC182BWHXL00250C – Item # EW-33684-07
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene(PE)
  • SizeX-Large
  • ColorWhite
  • SeamBound
  • ComplianceISO Class 4 (formerly Class 10)
  • DescriptionCoveralls, x-large, cleaned and individually wrapped
More About this Item
Tyvek® IsoClean® Coveralls are made using an exclusive flash spinning process. Excellent inherent barrier to dry particles, microorgainisms, and nonhazardous liquids. Coveralls feature zipper front, bound seams, and elastic wrists and ankles. White.
Compliance: ISO Class 5