DuPont Tyvek Coveralls with Elastic Cuffs and Hood, XX-Large
Representative image only.
DuPont Mfr # TY127SWH2X002500 – Item # EW-86225-24
Ideal for asbestos removal, general clean-up and maintenance, paint spraying, laboratories, and dry particulate protection
PRODUCT OPTIONS
$10.05USD / Each
BUY MORE & SAVE
  • 1-24$10.05USD / Each
  • 25+$9.05USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon;Polyester;Urethane;Carbon Fiber;Latex;Polyethylene;Polypropylene;Tyvek®;Vinyl;Cotton;Polyester / Cotton;Rubber;VR™;Neoprene;PVC
  • SizeXX-Large
  • ColorWhite
  • SeamSerged
  • ComplianceANSI/ISEA 101-1996
  • DescriptionCoveralls with Elastic Cuffs and Hood, XX-Large
More About this Item
Rugged Tyvek® fabric resists snags, tears, punctures, and abrasion. The nonwoven fabric provides protection against particulates. Tyvek is chemically and biologically inert, low linting, and static-free until laundered.
These white coveralls have a zippered front that makes them easy to put on and take off. Coveralls feature serged seams.
Compliance: Meets sizing requirements of ANSI/ISEA 101-1996
Features elastic wrists and ankles, and a hood.