DuPont Tyvek Coveralls with Elastic Cuffs, Hood and Boots; Large
Representative image only.
DuPont Mfr # TY122SWHLG002500 – Item # EW-86225-36
Ideal for asbestos removal, general clean-up and maintenance, paint spraying, laboratories, and dry particulate protection
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge
  • MaterialPolyethylene(PE)
  • ColorWhite
  • SeamSerged
  • ComplianceANSI/ISEA 101-1996
  • DescriptionCoveralls with Elastic Cuffs, Hood and Boots; Large
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Rugged Tyvek® fabric resists snags, tears, punctures, and abrasion. The nonwoven fabric provides protection against particulates. Tyvek is chemically and biologically inert, low linting, and static-free until laundered.
These white coveralls have a zippered front that makes them easy to put on and take off. Coveralls feature serged seams.
Compliance: Meets sizing requirements of ANSI/ISEA 101-1996
Features elastic wrists and ankles, a hood, and boots.