DWK Life Sciences (Kimble) 14395P-45 Replacement Pouring Ring for Media Bottles, PP, Clear; 10/CS
Representative image only.
DWK Life Sciences (Kimble) Mfr # 14395P-45 – Item # EW-34523-13
Find replacements or upgrade your bottle's caps and pouring rings
  • Ensure the bottle cap size is the same as the bottle neck size
  • Polypropylene is autoclavable, PBT and ETFE can withstand dry heat sterilization
  • Limited Lifetime Warranty
PRODUCT OPTIONS
$28.00USD / Case of 10
BUY MORE & SAVE
  • 1-4$28.00USD / Case of 10
  • 5-9$25.20USD / Case of 10
  • 10+$22.40USD / Case of 10
Often bought with…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Cap / Stopper SizeGL45
  • Cap ColorNatural
  • AutoclavableYes
  • DescriptionReplacement Pouring Ring for Media Bottles, PP, clear, 10/cs
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Ensure the bottle cap size is the same as the bottle neck size
  • Polypropylene is autoclavable, PBT and ETFE can withstand dry heat sterilization
  • Limited Lifetime Warranty