DWK Life Sciences (Kimble) Babcock pipettes; milk test, to contain; 12/cs
Representative image only.
DWK Life Sciences (Kimble) Mfr # 3005S-176 – Item # EW-34590-55
Get accurate results, easily
  • Conforms to AOAC Method 989.04
  • Features permanent brown-stain scales
PRODUCT OPTIONS
$336.00USD / Case of 12
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)17600
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)17.6
  • Max Dispensing (mL)17.6
  • Accuracy (%)±0.050mL
  • DescriptionBabcock pipettes; milk test, to contain; 12/cs
Key Features
  • Conforms to AOAC Method 989.04
  • Features permanent brown-stain scales
More About this Item
Model 34590-50 is designed to comply with the Pennsylvania modified Babcock as referenced in APHA 15.8. Models 34590-55,-56,-60, and -61 include a special rubber washer that allows air displaced by incoming milk to escape freely. Models 34590-55 and -56 meet ASTM Specification E1043, Type II A requirements; models 34590-60 and -61 meet ASTM Specification E1043, Type II B requirements. All pipettes feature permanent brown-stain scales.