DWK Life Sciences (Kimble) Glass Desiccator with Plastic top Knob, 150 mm

Representative image only.
DWK Life Sciences (Kimble) Mfr # 31050-150 – Item # EW-06536-41
Durable and efficient for daily use
  • Conforms to USP Type III and ASTM E438, Type II requirements

PRODUCT OPTIONS

$433.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Plate Size (mm)150
  • Exterior Height (InchShort)10
  • Exterior Height (cm)25.5
  • MaterialGlass
  • DescriptionGlass Desiccator with Plastic top Knob, 150 mm

Key Features

  • Conforms to USP Type III and ASTM E438, Type II requirements

More About this Item

Soda-lime glass desiccators include a metal plate, s solid lid with a threaded plastic top knob and a recessed base for containment of desiccant. Excellent for storage of hygroscopic chemicals, and to prevent absorption of moisture when using crucibles, weighing bottles, and watch glasses.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat