DWK Life Sciences (Kimble) HPLC Mobile Phase Reservoir, Glass, Conical Bottom; 1.0 Liter
Representative image only.
DWK Life Sciences (Kimble) Mfr # 953901-1002 – Item # EW-02017-05
$351.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Bottom TypeConical
  • Total Volume Capacity (mL)1000
  • DescriptionHPLC Mobile Phase Reservoir, Glass, Conical Bottom; 1.0 Liter
More About this Item
These reservoirs are manufactured of high-quality borosilicate glass for maximum chemical resistance. Reservoirs are coated with plastic for safety and UV resistance. Select from conical bottom configuration for maximum solvent utilization or flat bottom reservoirs.