DWK Life Sciences (Kimble) Kimax Friedrich Condenser; 125 mL; 1/Pk
Representative image only.
DWK Life Sciences (Kimble) Mfr # 456250-0021 – Item # EW-99037-32
Choose the condenser by type and sample volume
  • Allihn condenser (aka bulb or reflux condenser) better suited to basic laboratory applications
  • 1 year warranty
PRODUCT OPTIONS
$324.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)125
  • Condenser TypeFreidrich
  • Flask Joint Size34/45
  • Condenser Joint Size34/45
  • DescriptionFriedrich Condenser; 125 mL; 1/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Allihn condenser (aka bulb or reflux condenser) better suited to basic laboratory applications
  • 1 year warranty
More About this Item
Freidrichs condenser (aka spiral cold finger condenser) better suited to heavy or difficult applications.