DWK Life Sciences (Kimble) Kimax Soxhlet Round Bottom Flask; 300 mL; 1/Pk
Representative image only.
DWK Life Sciences (Kimble) Mfr # 601000-0524 – Item # EW-99037-64
Choose an extraction flask by volume and bottom shape
  • Flat bottom flasks sit easily on flat surfaces
PRODUCT OPTIONS
$54.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)300
  • Flask TypeRound-bottom
  • Flask Joint Size24/40
  • Condenser Joint Size24/40
  • DescriptionSoxhlet Round Bottom Flask; 300 mL; 1/Pk
Key Features
  • Flat bottom flasks sit easily on flat surfaces
More About this Item
Round bottom flasks require a cork or rubber ring to stand, or a neck clamp attached to a laboratory support rod