DWK Life Sciences (Kimble) Silicone Lid, Full Set; Cyan
Representative image only.
DWK Life Sciences (Kimble) Mfr # 291110029 – Item # EW-34500-31
Versatile lids work on various vessel diameters and with spouts, beads, or thread finishes
  • Constructed of stretchable, chemically resistant, and heat-resistant silicone
  • Quickly identify or track samples visually with different color options
  • Reusable dishwasher-safe lids reduce environmental impact and lowers costs
  • Full set includes one small lid (43 to 61 mm diameter), one medium lid (64 to 76 mm diameter) and one large lid (84 to 116 mm diameter)
PRODUCT OPTIONS
$9.00USD / Pkg of 3
OFTEN BOUGHT WITH…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSilicone
  • Cap MaterialSilicone
  • Cap / Stopper SizeSmall, Medium, and Large
  • Cap ColorCyan
  • AutoclavableYes
  • Special FeaturesOne 43 to 61 mm diameter; One 64 to 76 mm diameter; One 84 to 116 mm diameter
  • Withstands High Temperature (° F)346
  • Withstands Low Temperature (° F)-40
  • DescriptionSilicone Lid, Full Set; Cyan
Includes
one small cyan lid, one medium cyan lid, and one large cyan lid.
Key Features
  • Constructed of stretchable, chemically resistant, and heat-resistant silicone
  • Quickly identify or track samples visually with different color options
  • Reusable dishwasher-safe lids reduce environmental impact and lowers costs
  • Full set includes one small lid (43 to 61 mm diameter), one medium lid (64 to 76 mm diameter) and one large lid (84 to 116 mm diameter)
More About this Item
These DWK KIMBLE® silicone lids are made of stretchable silicone to fit on beakers, flasks, bottles, and cylinders of various diameter vessels. The stretchable yet secure lids provide a tight fit on vessels with spouts, beads, and threaded finishes. This versatility allows for many uses beyond the limits of traditional films and covers. Lids are available in three colors for quick visual identification—pink, cyan, and green. The stretchable silicone material is compatible with lab markers to allow for optimal sample identification.