DWK Life Sciences (Wheaton) 356887 Cell Culture Spin Flask, 3-Liter; 1/Pk
Representative image only.
DWK Life Sciences (Wheaton) Mfr # 356887 – Item # EW-29300-04
Perfect for small scale operations, or quick tests of your fermentation process
  • Designed for microcarrier and suspension cultures, such as insect cells, hybridomas, and adapted cell lines
  • Bottom dimple prevents cell crushing and improves circulation
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)3000
  • Overall Height (cm)34.1
  • DescriptionCell Culture Spin Flask, 3-Liter; 1/Pk
Key Features
  • Designed for microcarrier and suspension cultures, such as insect cells, hybridomas, and adapted cell lines
  • Bottom dimple prevents cell crushing and improves circulation
More About this Item
Use the 45-mm sidearms as air vents, inoculation ports, or media inlet/outlet ports, or use sidearms to insert probes for pH, dissolved oxygen, or CO2 measurement and control. Flasks are made of borosilicate glass, and include a PTFE and glass impeller assembly. Sidearms have PTFE-faced silicone liners. Top (center) cap has two liners: one silicone and one PTFE (to be used together). Entire spin flask unit is autoclavable. Additional sizes are available; please call our Applications Specialists for details.