DWK Life Sciences (Wheaton) 844026 Clear Glass Safety Bottle w/ Side-Arm, 500 mL, GL32/GL16, 33 mm cap
Representative image only.
DWK Life Sciences (Wheaton) Mfr # 844026 – Item # EW-07914-82
  • 3 Month Product / 12 Month Parts and Labor Warranty
PRODUCT OPTIONS
$245.50USD / Each
BUY MORE & SAVE
  • 1-4$245.50USD / Each
  • 5-9$220.95USD / Each
  • 10+$196.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)500
  • Capacity (oz)16
  • MaterialGlass
  • Cap / Stopper Size33 mm
  • Bottle ColorClear
  • DescriptionClear Glass Safety Bottle w/ Side-Arm, 500 mL, GL32/GL16, 33 mm cap
  • Warranty3 Month Product / 12 Month Parts and Labor
Key Features
  • 3 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Plain amber bottles (USP type 3) are available in 250-, 1000-, and 2500-mL sizes. PVC-coated amber bottle 07914-84 is designed for safety—won’t shatter like regular bottles. Clear, borosilicate glass bottle 07914-82 features a sidearm for a 0.2-µm air filter.