DWK Life Sciences (Wheaton) CompletePAK® Sterile Serum Vial, 10 mL; 158/Cs

Representative image only.
DWK Life Sciences (Wheaton) Mfr # W987002-S – Item # EW-08918-81
Order individual system components—pre-sterilized!
  • Keep a spare or order components separately to buil your own system
  • Sterile, clear-glass vial, 10 mL capacity

PRODUCT OPTIONS

$589.00USD / Case of 158

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)10
  • Tube MaterialGlass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeNot applicable
  • Cap Diameter (mm)20
  • Cap colorNot applicable
  • SterileYes
  • AutoclavableYes
  • DescriptionCompletePAK® Sterile Serum Vial, 10 mL; 158/Cs
Sterile

Key Features

  • Keep a spare or order components separately to buil your own system
  • Sterile, clear-glass vial, 10 mL capacity

More About this Item

Separate vial components (vials, stoppers, and seals) come in pre-sterilized cases. Order a spare, or build-your-own vial system.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat