Dwyer 166T Pitot Tubetelescoping S.S. Adjustabl 11.5in To 36in

Representative image only.
Dwyer Mfr # 166T – Item # EW-10778-98
Get better positioning in airstream
  • A unique airflow sensor
  • Can be adjusted for any duct insertion length from 11.5" to 36"
  • Replace up to five conventional fixed-length Pitot tubes
  • Securely protected by a custom-fitted polyethylene carrying case
  • Stainless steel construction resists corrosion
  • Hemispherical tip has 1.0 flow coefficient
  • 1⁄8" diameter (3.2 mm); largest section is 3⁄8" (9.5 mm)
  • 1 year warranty

$355.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Probe length (in)36
  • DescriptionPitot Tubetelescoping S.S. Adjustabl 11.5in To 36in
  • Warranty1 year
1 Year Warranty

Key Features

  • A unique airflow sensor
  • Can be adjusted for any duct insertion length from 11.5" to 36"
  • Replace up to five conventional fixed-length Pitot tubes
  • Securely protected by a custom-fitted polyethylene carrying case
  • Stainless steel construction resists corrosion
  • Hemispherical tip has 1.0 flow coefficient
  • 1⁄8" diameter (3.2 mm); largest section is 3⁄8" (9.5 mm)
  • 1 year warranty

More About this Item

Telescoping sections lock in place as they are extended, enabling use of the handle grip to gauge proper alignment of the tip within the airstream.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat